missing translation for 'onlineSavingsMsg'
Learn More

Promega Wizard™ Magnetic DNA Purification System for Food

Catalog No. PRFF3750
Encompass
Change view
Click to view available options
No. of Reactions:
200 Preps
400 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRFF3750 200 Preps
PRFF3751 400 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRFF3750 Supplier Promega Supplier No. FF3750
Only null left

Includes

MagneSil Paramagnetic Particles; Lysis Buffer A, Food; Lysis Buffer B, Food; Precipitation Solution, Food; RNase A Solution

Requires

MagneSphere Technology Magnetic Separation Stand (Z5332 or Z5342)

The Wizard™ Magnetic DNA Purification System for Food is designed for purification of DNA from a variety of food samples.

  • Rapid Isolation of DNA from Many Food Types
  • Requires minimal centrifugation and eliminates organic extractions
  • Purified DNA suitable for PCR-based GMO testing

U.S. Patents 6,027,945 and 6,368,800, Australian Patent 732756 and other patents and patents pending.

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Purifies DNA from a variety of food samples including corn seeds, cornmeal, soybeans, Soy flour and Soy milk
Includes 100mL Lysis Buffer A, 100mL Lysis Buffer B, 1mL RNase A, 19mL Magnesil™ paramagnetic particles, 150mL precipitation solution
No. of Reactions 200 Preps
Product Type System