Learn More
Thermo Scientific™ Wide-Mouth Short-Profile Clear Glass Jars with Closure
GREENER_CHOICE
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes with these wide-mouth, short-profile clear glass jars with closure when light protection is not required.
$208.41 - $441.26
Specifications
Color Clear
Description Clear Glass Short Jar
Material Glass (Type 3)
Closure Color White
Liner Material PTFE
View More Specs
Products 14
Catalog Number Mfr. No. Capacity (Metric) Level Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Level Closure Size Price Quantity & Availability  
05719258
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
120 0060
60 mL Unprocessed 53-400 mm
Case of 24 Each for $304.15
Only null left
05719257
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
320 0060
60 mL Certified 53-400 mm
Case of 24 Each for $403.88
Only null left
0571951
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
220 0060
60 mL Processed 53-400 mm
Case of 24 Each for $349.03
Only null left
0571957
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
320 0125
125 mL Certified 58-400 mm
Case of 24 Each for $441.26
Only null left
0571955
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
220 0125
125 mL Processed 58-400 mm
Case of 24 Each for $391.41
Only null left
0571953
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
120 0125
125 mL Unprocessed 58-400 mm
Case of 24 Each for $331.59
Only null left
0571965
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
320 0250
250 mL Certified 70-400 mm
Case of 12 Each for $244.82
Only null left
0571959
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
120 0250
250 mL Unprocessed 70-400 mm
Case of 12 Each for $208.41
Only null left
0571961
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
220 0250
250 mL Processed 70-400 mm
Case of 12 Each for $216.65
Only null left
0571976
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
320 0500
500 mL Certified 89-400 mm
Case of 12 Each for $266.76
Only null left
0571972
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
220 0500
500 mL Processed 89-400 mm
Case of 12 Each for $232.85
Only null left
05719275
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
320 2000
2000 mL Certified 110-400 mm
Case of 6 Each for $309.15
Only null left
05719277
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
120 2000
2000 mL Unprocessed 110-400 mm
Case of 6 Each for $234.59
Only null left
05719276
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
220 2000
2000 mL Processed 110-400 mm
Case of 6 Each for $276.74
Only null left
Description
Description
Collect soil, sludge and waste samples for a variety of organic and inorganic target analytes with Thermo Scientific™ Wide-Mouth Short-Profile Clear Glass Jars when light protection is not required. Jars are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses, Processed the same way as the certified jars but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Clear Type III soda-lime glass jar
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 60 to 2000mL
  • Closures assembled on bottles
Includes:
Certified jars come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each jar individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed jars are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded, and CofA is not included.
Unprocessed jars are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.
Specifications
Specifications
Clear
Glass (Type 3)
PTFE
Jar
Closed-top Cap
Clear Glass Short Jar
White
Wide
Polypropylene