missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Wide Mouth Packer Bottles, Amber, Safety Coated, PV-Lined Lids
Safety-coated wide mouth packer bottles made of Type III amber soda-lime glass with white polypropylene polyvinyl-lined caps.
$283.36
Specifications
| Color | Amber |
|---|---|
| Material | Borosilicate Glass (Type 3) |
| Closure Color | White |
| Surface Treatment | Safety Coated |
| Closure Included | Yes |
Description
- Made from amber soda-lime glass that conforms to USP Type III requirements
- Amber bottles protect light sensitive samples or dry powders during storage
- Wide mouth opening for easy removal of contents
Specifications
| Amber | |
| White | |
| Yes | |
| Straight-sided Jar | |
| Polypropylene |
| Borosilicate Glass (Type 3) | |
| Safety Coated | |
| Wide | |
| Polyvinyl |