missing translation for 'onlineSavingsMsg'
Learn More

White Swan™ Essentials™ Women's Princess-Style Lab Coats

Knee-length lab coats feature long set-in sleeves and four convenient pockets.

$41.73 - $59.81

Specifications

Product Type General Purpose Lab Coat
Gender Women's
Material Polyester, Cotton
Color White
Closure Type Buttons
View More Specs

Products 16
Catalog Number Mfr. No. Size Size (English) Waist Size (Metric) Chest Size (Metric) Hip Price Quantity  
Catalog Number Mfr. No. Size Size (English) Waist Size (Metric) Chest Size (Metric) Hip Price Quantity  
19-154-101
View Documents
White Swan™
47401W06
6 26 in. 86.36 cm 92.71 cm
Each for $49.25
Only null left
 
19-154-102
View Documents
White Swan™
47401W08
8 27 in. 88.9 cm 95.25 cm
Each for $41.73
Only null left
 
19-154-103
View Documents
White Swan™
47401W10
10 28 in. 91.44 cm 97.79 cm
Each for $41.73
Only null left
 
19-154-104
View Documents
White Swan™
47401W12
12 29.5 in. 95.25 cm 101.6 cm
Each for $49.25
Only null left
 
19-154-105
View Documents
White Swan™
47401W14
14 31 in. 99.06 cm 105.4 cm
Each for $49.25
Only null left
 
19-154-100
View Documents
White Swan™
47401W04
4 25 in. 83.82 cm 90.17 cm
Each for $49.25
Only null left
 
19-154-106
View Documents
White Swan™
47401W16
16 32.5 in. 102.87 cm 109.22 cm
Each for $41.73
Only null left
 
19-154-107
View Documents
White Swan™
47401W18
18 34.5 in. 108 cm 114.3 cm
Each for $49.25
Only null left
 
19-154-109
View Documents
White Swan™
47401W40
Medium 38.5 in. 118 cm 124.46 cm
Each for $59.81
Only null left
 
19-154-111
View Documents
White Swan™
47401W44
X-Large 127 cm 134.62 cm
Each for $59.81
Only null left
 
19-154-108
View Documents
White Swan™
47401W20
20 113 cm 119.4 cm
Each for $49.25
Only null left
 
19-154-113
View Documents
White Swan™
47401W48
3X-Large 46.5 in. 137.16 cm 144.78 cm
Each for $59.81
Only null left
 
19-154-110
View Documents
White Swan™
47401W42
Large 40.5 in. 122 cm 129.54 cm
Each for $59.81
Only null left
 
19-154-112
View Documents
White Swan™
47401W46
2X-Large 132.08 cm 139.7 cm
Each for $50.68
Only null left
 
19-154-114
View Documents
White Swan™
47401W50
4X-Large 142.24 cm 149.86 cm
Each for $59.81
Only null left
 
19-154-115
View Documents
White Swan™
47401W52
5X-Large 50.5 in. 147.32 cm 154.94 cm
Each for $59.81
Only null left
 
Description

Description

  • Composition: 80% Polyester, 20% Cotton
  • Women's princess cut
  • Button closure
  • Stain release finish, non-sterile
Specifications

Specifications

General Purpose Lab Coat
Polyester, Cotton
Buttons
Non-sterile
4
Set Sleeve
39.5 in.
Women's
White
Reusable
Princess
Stain Release
Mock belt, single breasted, 5-button front
100.3 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'