Learn More
DWK Life Sciences Wheaton™ Wide-Mouth Bottles with Ground Glass Stoppers
Borosilicate glass bottle and stopper features no-drip pour lip. Ground glass stopper fits securely to prevent contamination
$634.33 - $2,282.56
Specifications
Color Clear
Material Borosilicate Glass
Closure Type Stopper
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
02911308
View Documents Promotion Details
DWK Life Sciences
W216015
100 mL
Case of 6 Each for $634.33
Only null left
02910G
View Documents Promotion Details
DWK Life Sciences
216019
500 mL
Case of 10 Each for $1,579.94
Only null left
02910H
View Documents Promotion Details
DWK Life Sciences
DWK216020
1000 mL
Case of 10 Each for $2,282.56
Only null left
Description
Description
  • Wide mouth ideal for solids and viscous liquids
  • Easy to clean
  • Autoclavable
Specifications
Specifications
Clear
Borosilicate Glass
Stopper