missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ V Vial™ with Crimp Top Finish; no Closures

Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.

$217.43 - $451.15

Specifications

Color Clear
Material Borosilicate Glass (Low Extractable)
For Use With (Application) Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Graduated No
Style Conical V
View More Specs

Products 6
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Diameter (Metric) Outer Price Quantity & Availability  
22021827
View Documents Promotion Details
DWK Life Sciences
W986213NG
0.3 mL 32 mm 13 mm
Case of 12 Each for $245.07
Only null left
 
22021829
View Documents Promotion Details
DWK Life Sciences
W986216NG
2 mL 40 mm 20 mm
Case of 12 Each for $289.04
Only null left
 
22021832
View Documents Promotion Details
DWK Life Sciences
W986219NG
5 mL 61 mm 20 mm
Case of 12 Each for $289.98
Only null left
 
0334115
View Documents Promotion Details
DWK Life Sciences
W986211NG
0.1 mL 32 mm 12 mm
Case of 12 Each for $217.43
Only null left
 
03410028
View Documents Promotion Details
DWK Life Sciences
W986214NG
1 mL 41 mm 13 mm
Case of 12 Each for $234.73
Only null left
 
03410036
View Documents Promotion Details
DWK Life Sciences
986217NG
3 mL 46 mm 20 mm
Case of 12 Each for $451.15
Only null left
 
Description

Description

Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.

  • Not graduated
  • Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
  • Conical interior bottoms are designed to provide total downward drainage for maximum sample retrieval

Order Info

Closures are sold separately.

Specifications

Specifications

Clear
Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Conical V
ASTM E438, USP Type I, Type I, Class A
Borosilicate Glass (Low Extractable)
No
Crimp Cap
Videos
SDS
Documents

Documents