Learn More
DWK Life Sciences Wheaton™ V Vial™ with Crimp Top Finish; no Closures
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
Supplier:  DWK Life Sciences W986211NG
 View more versions of this product
Catalog No. 0334115
Only null left
Description
Description
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
  • Not graduated
  • Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
  • Conical interior bottoms are designed to provide total downward drainage for maximum sample retrieval
Order Info
Closures are sold separately.
Specifications
Chemical Identifiers
Clear
Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Conical V
ASTM E438, USP Type I, Type I, Class A
Borosilicate Glass (Low Extractable)
No
Crimp Cap
Specifications
0.196 in.
Clear
Borosilicate Glass (Low Extractable)
Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Conical V
Crimp Cap
32 mm
5 mm
12 mm
0.1 mL
No
0.027 dr.
ASTM E438, USP Type I, Type I, Class A
Product Suggestions
Product Suggestions