Learn More
DWK Life Sciences Wheaton™ Staining Jars and Accessories
Staining Jars and Accessories
$150.79 - $840.54
| Catalog Number | Mfr. No. | Type | Slide Count | Material | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Slide Count | Material | Price | Quantity & Availability | |||
|
088131
|
DWK Life Sciences
900575 |
Slide Grip | 5 | Polypropylene |
Case of 2 Each for $150.79
|
|
|||
|
22312143
|
DWK Life Sciences
900470 |
Coplin Staining Jar | 5 Vertically, 10 Back to Back | Soda Lime Glass |
Pack of 6 Each for $840.54
|
|
|||
Description
Coplin Staining Jar:
- Made from soda-lime glass
- 60mL capacity
- Allows for easy and safe transfer of five slides
- Fits into Wheaton Coplin staining jars
Specifications
| Staining |