missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Ideal for soil sampling and environmental applications
$172.82 - $282.46
Specifications
| Color | Clear |
|---|---|
| Material | Soda Lime Glass |
| Closure Color | White |
| Closure Liner | PTFE |
| Closure Material | Polypropylene |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|
02911411
|
DWK Life Sciences
W216908 |
60 mL | 53-400 mm |
Case of 24 Each for $181.75
|
|
||||
|
02911412
|
DWK Life Sciences
W216909 |
125 mL | 58-400 mm |
Case of 24 Each for $221.71
|
|
||||
|
02911413
|
DWK Life Sciences
W216910 |
250 mL | 70-400 mm |
Case of 12 Each for $172.82
|
|
||||
|
02911414
|
DWK Life Sciences
W216911 |
500 mL | 89-400 mm |
Case of 12 Each for $262.89
|
|
||||
|
02911415
|
DWK Life Sciences
W216912 |
1000 mL | 89-400 mm |
Case of 12 Each for $282.46
|
|
||||
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Walls allow for complete removal of contents
- Wide-mouth design is great for large solid samples
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear USP Type III soda-lime glass
Specifications
| Clear | |
| White | |
| Polypropylene |
| Soda Lime Glass | |
| PTFE | |
| Screw Cap |