missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
$277.50 - $682.20
Specifications
| Color | Black |
|---|---|
| For Use With (Application) | Ideal for cell culture work |
| Material | Phenolic |
| Liner Material | Styrene Butadiene Rubber |
| Product Type | Screw Cap |
| Catalog Number | Mfr. No. | Closure Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Closure Size | Price | Quantity | |||||
|
02-912-245
|
DWK Life Sciences
239298 |
20-400 |
N/A
|
|
|||||
|
03-084-120
|
DWK Life Sciences
240215 |
18-400 |
N/A
|
|
|||||
|
02-912-247
|
DWK Life Sciences
239300 |
24-400 |
N/A
|
|
|||||
Description
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
- 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
- Aluminum foil faced pulp board liners can be used with organic solvents
- Use LDPE lined caps with distilled water, analytical standards and reagents
Specifications
| Black | |
| Phenolic | |
| Screw Cap |
| Ideal for cell culture work | |
| Styrene Butadiene Rubber | |
| Autoclavable |