Learn More
DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
Supplier:  DWK Life Sciences 239300
 View more versions of this product
Catalog No. 02-912-247
Only null left
Explore available promotions
Description
Description
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
  • 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
  • Aluminum foil faced pulp board liners can be used with organic solvents
  • Use LDPE lined caps with distilled water, analytical standards and reagents
Specifications
Specifications
Black
Phenolic
Screw Cap
Autoclavable
Ideal for cell culture work
Styrene Butadiene Rubber
24-400
Product Suggestions
Product Suggestions