missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps
Description
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
- 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
- Aluminum foil faced pulp board liners can be used with organic solvents
- Use LDPE lined caps with distilled water, analytical standards and reagents
Specifications
Specifications
| Color | Black |
| For Use With (Application) | Ideal for cell culture work |
| Material | Phenolic |
| Liner Material | Styrene Butadiene Rubber |
| Product Type | Screw Cap |
| Closure Size | 20-400 |
| Autoclavable | Autoclavable |