missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD
Offers high filtration efficiency and excellent filtrate purity
Supplier: Cytiva 2813T
Description
- Allows usage with broad range of solutions, pH, and temperature
- High flow and high retention capacity
- Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
- Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
Chemical Identifiers
| Polycap HD 150 | |
| 5/Pk. |
Specifications
| Polycap HD 150 Capsule Filter, 10.0 μm, with FNPT inlet and outlet (5 pcs) | |
| Polycap HD 150 | |
| With Female NPT inlet/outlet | |
| 5/Pk. |
| 10 μm | |
| Polypropylene | |
| 4.1 bar (60 psi) |