missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD

Offers high filtration efficiency and excellent filtrate purity

Supplier:  Cytiva 2814T

 View more versions of this product

Catalog No. 09927684


Only null left

Description

Description

  • Allows usage with broad range of solutions, pH, and temperature
  • High flow and high retention capacity
  • Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port

Specifications

Chemical Identifiers

Polycap HD 150
5/Pk.

Specifications

Polycap HD 150 Capsule Filter, 20.0 μm, with FNPT inlet and outlet (5 pcs)
Polycap HD 150
With Female NPT inlet/outlet
5/Pk.
20 μm
Polypropylene
4.1 bar (60 psi)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions