missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 36 AS
Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.
$112.48 - $779.58
Specifications
| Material (Membrane) | Nylon |
|---|---|
| Filtration Area | 400 cm2 |
| For Use With (Application) | Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. |
| Housing Material | Polypropylene |
| Max. Pressure | 4.1 bar (60 psi) |
| Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Sterility | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Sterility | Price | Quantity | ||
|
09-743-90
|
Cytiva
67083602 |
0.2 μm | 1/2 SB inlet | 1/2 SB outlet | Sterile |
Each for $112.48
|
|
||
|
09-743-94
|
Cytiva
67093602 |
0.2 μm | MNPT inlet | SB outlet | Sterile |
Each for $119.30
|
|
||
|
09-744-48
|
Cytiva
67053604 |
0.45 μm | SB inlet | SB outlet | Sterile |
Each for $122.88
|
|
||
|
09-744-47
|
Cytiva
67053602 |
0.2 μm | SB inlet | SB outlet | Sterile |
Each for $132.45
|
|
||
|
09-744-61
|
Cytiva
67063602 |
0.2 μm | SB inlet | SB outlet plus filling bell | Sterile |
Each for $133.98
|
|
||
|
09927913
|
Cytiva
2606T |
0.2 μm | FNPT | FNPT | Non-sterile |
Pack of 5 Each for $779.58
|
|
||
Description
Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.- First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
- Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
- Housing thermally fused (no glues, adhesives, or extraneous materials).
- Integrity-testable by bubble point, pressure decay, or forward flow methods.
- Provides highly effective filtration area in a small size.
- Autoclavable; some presterilized with gamma irradiation.
- Manufactured in clean room facilities under ISO Quality Systems.
Specifications
| Nylon | |
| Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. | |
| 4.1 bar (60 psi) |
| 400 cm2 | |
| Polypropylene |