missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Whatman™ Polycap Disposable Capsules, 36 AS

Catalog No. 09927913
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
1/2 SB inlet
FNPT
MNPT inlet
SB inlet
Outlet Style:
1/2 SB outlet
FNPT
SB outlet
SB outlet plus filling bell
Sterility:
Non-sterile
Sterile
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Inlet Style Outlet Style Sterility
09927913 0.2 μm FNPT FNPT Non-sterile
0974390 0.2 μm 1/2 SB inlet 1/2 SB outlet Sterile
0974394 0.2 μm MNPT inlet SB outlet Sterile
0974448 0.45 μm SB inlet SB outlet Sterile
0974447 0.2 μm SB inlet SB outlet Sterile
0974461 0.2 μm SB inlet SB outlet plus filling bell Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09927913 Supplier Cytiva™ Supplier No. 2606T
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.

Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
  • Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
  • Housing thermally fused (no glues, adhesives, or extraneous materials).
  • Integrity-testable by bubble point, pressure decay, or forward flow methods.
  • Provides highly effective filtration area in a small size.
  • Autoclavable; some presterilized with gamma irradiation.
  • Manufactured in clean room facilities under ISO Quality Systems.

Specifications

Description Polycap AS 36 Capsule Filter, 0.2 μm, FNPT inlet and outlet (5 pcs)
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 400 cm2
For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing Material Polypropylene
Inlet Style FNPT
Max. Pressure 4.1 bar (60 psi)
Outlet Style FNPT