missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva™ Whatman™ Nylon Membranes
Description
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
- Pure nylon membranes
- Tear-resistant
- Filter many organic solvents
- Autoclavable up to 135°C
- Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
- Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Nylon |
| Pore Size | 0.2 μm |
| For Use With (Application) | For filtering aqueous solutions and most organic solvents |
| Hydrophobicity | Hydrophilic |
| Quantity | 50/Pk. |
| Diameter (Metric) | 90 mm |