missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Nylon Membranes

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

Supplier:  Cytiva 7404002

 View more versions of this product

Catalog No. 09-902-9

Only null left
Explore available promotions

Description

Description

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

 

  • Pure nylon membranes
  • Tear-resistant
  • Filter many organic solvents
  • Autoclavable up to 135°C
  • Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
  • Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing

 

Specifications

Specifications

Autoclavable
0.45 μm
100/Pk.
Nylon
Hydrophilic
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions