missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Nylon Membranes

Catalog No. 099025
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.5 μm
0.8 μm
Diameter (Metric):
13 mm
25 mm
47 mm
90 mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric)
099025 0.2 μm 47 mm
0991032B 0.2 μm 90 mm
0990213 0.8 μm 47 mm
099028 0.5 μm 13 mm
0990210 0.45 μm 47 mm
099023 0.2 μm 13 mm
099029 0.45 μm 25 mm
0991033 0.45 μm 90 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 099025 Supplier Cytiva Supplier No. 7402004
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

 

  • Pure nylon membranes
  • Tear-resistant
  • Filter many organic solvents
  • Autoclavable up to 135°C
  • Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
  • Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing

 

Specifications

Autoclavable Autoclavable
Filter Type Membrane Filter
Material Nylon
Pore Size 0.2 μm
Hydrophobicity Hydrophilic
Quantity 100/Pk.
Type Filtration Membrane Disc
Diameter (Metric) 47 mm