missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Nylon Membranes
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
Supplier: Cytiva 7408004
Description
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
- Pure nylon membranes
- Tear-resistant
- Filter many organic solvents
- Autoclavable up to 135°C
- Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
- Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing
Specifications
| Autoclavable | |
| Nylon | |
| Hydrophilic | |
| Filtration Membrane Disc |
| Membrane Filter | |
| 0.8 μm | |
| 100/Pk. | |
| 47 mm |