Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes
Color contrast helps easier particle detection.
Supplier: Cytiva 10407312
Description
- Nontoxic grids do not inhibit bacterial growth
- Composed of cellulose nitrate and cellulose acetate
- High degree of internal surface area for greater adsorption of product
- High dirt-loading capacity
- Biologically inert with good thermal stability
- No surfactants to contaminate samples
- Uniform microporous structure of membrane allows high flow rates
- Sterile allows critical applications
Specifications
| Autoclavable | |
| MCE | |
| Sterile | |
| Membrane |
| White | |
| 0.45 μm | |
| Black |