missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 0.45 μm Pore Size (ME 25) Mixed Cellulose Ester Membranes, White/Black Grid
Whatman™ (ME 25) Mixed Cellulose Ester Membranes, White/Black Grid are particularly effective in applications requiring higher flow rates and larger volume filtration. Supplied with clearly defined grid lines at regularly spaced intervals to assist in microbiological colony counting.
Supplier: Cytiva 10406572
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 25
- Pore Size: 0.45 μm
- Size (Metric) Grid: 5 mm
- Color: White
- Grid Color: Black
- Autoclavable: Yes
- Sterility: Sterile
- Diameter (Metric): 50 mm
- Flow Rate: 12.5 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
| Autoclavable | |
| Gridded Filter | |
| ME 25 | |
| Sterile | |
| Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
| 1.97 in. |
| White | |
| MCE | |
| 0.45 μm | |
| 12.5 sec./100 mL/12.5 cm² (at Δp = 0.9 bar) | |
| Black | |
| 50 mm |