missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA PlantSaver

Catalog No. 09924590
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09924590 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09924590 Supplier QIAGEN Supplier No. WB120065
Only null left

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

The 4 spot area QIAcard FTA PlantSaver format provides a laminated flap that allows you to vigorously crush plant or insect samples into the FTA matrix without damaging the FTA card.

Features:

  • Simply pressing plant samples onto the FTA matrix or applying homogenate captures and stabilizes the DNA.
  • Store and collect at room and high temperatures for convenient collection in the field.
  • Smaller sample size: Only young leaves are required, reducing growing time and speeding research.
  • Fast purification in 30 minutes: PCR-ready DNA can be purified on FTA in three simple steps, all in a single tube at room temperature.

Applications:

  • Analyze plant DNA by PCR-based assays
  • Detection of Genetically Modified (GM) organisms
  • Marker selected breeding
  • Varietal identification
  • Phylogeny analysis
  • Amplification of low copy number loci
  • Invader™ assays
  • Multiple displacement amplification
  • Transgenic identification

Analysis:

  • Plant tissue is physically crushed on the card and the DNA binds to the matrix.
  • The chemical coating on the FTA card protects the DNA from degradation and allows the cards to be stored at room temperature for extended periods of time.
  • To prepare the sample for PCR, the matrix is washed with two nontoxic aqueous buffers.
  • The DNA remains bound to the matrix throughout purification and a small disk (1.2 or 2 mm in diameter) of the matrix provides enough template for PCR analysis.

Specifications

Description FTA PlantSaver card with 4 sample areas per card
Format Card
Sample Type Plants, Biological
Formulation Indicating
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 4
Product Type FTA PlantSaver Card
Quantity 100 Pk.