missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Micro
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Micro with 1 sample area per card.
- QIAcard FTA Micro format with 1 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Specifications
| Description | Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Micro Cards. |
| Sample Type | Blood |
| For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
| No. of Samples | 1 |
| Product Type | FTA Micro Card |
| Quantity | 25 Pk. |
| Sample Volume | 125 μL |