Learn More
QIAGEN QIAcard FTA Gene
For collection, stabilization, processing, transport and archiving of biological samples. 3 sample areas per card.
Supplier:  QIAGEN WB120208
Catalog No. 09-923-346
Only null left
Explore available promotions
Description
Description
For collection, stabilization, processing, transport and archiving of biological samples. 3 sample areas per card.
  • Three sample areas in a chipboard frame suitable for several automated dispensing/pipetting or punching systems.
  • Storage of up to 75 μL of blood (whole blood, EDTA, sodium citrate, ACD, or heparin anticoagulated blood) to each printed circle of each FTA Gene Card (total 225 μL per card).
  • Apply only one blood sample to each card.
Specifications
Specifications
FTA Gene Card, 3 sample areas per card, 75 μl max volume/sample area, 225 μl max total volume/card
3
100 Pk.
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
FTA Gene Card
75 μL
Product Suggestions
Product Suggestions