missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA DMPK

Catalog No. 09801000
Change view
Click to view available options
Product Type:
QIAcard FTA DMPK-A
QIAcard FTA DMPK-B
QIAcard FTA DMPK-C
Formulation:
Type A
Type B
Type C
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Formulation
09801000 QIAcard FTA DMPK-C Type C
09800998 QIAcard FTA DMPK-A Type A
09800999 QIAcard FTA DMPK-B Type B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09801000 Supplier QIAGEN Supplier No. WB129243

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

QIAcard FTA DMPK Cards for Drug Metabolism Pharmacokinetic enable microvolume sampling with only 10 to 20 μL per sample and are designed for use with blood samples. The cards can also be used with colorless samples such as plasma, cerebrospinal fluid and urine.

QIAcard FTA DMPK-A and QIAcard FTA DMPK-B contains impregnated chemicals enabling protein denaturation to inactivate endogenous enzymes. Cell lysis releases endogenous cellular materials onto card. Stabilization of DNA allows resampling of blood spot for pharmacogenomics.Impregnated chemicals may interfere with mass spectrometry detection (e.g. ion suppression).

QIAcard FTA DMPK-C contain pure cellulose. No impregnated chemicals to interfere with analysis. Proteins are not denatured so cards may be better suited for protein based biomolecules and antibody studies.

Analysis:

Analysis primarily with HPLC or UHPLC followed by MS/MS detection, direct desorption MS techniques such as DESI. Methanol or aqueous/organic mixtures have been shown to work well for small molecules and aqueous buffers are recommended for proteins.

Select a good solvent for the analyte while maintaining compatibility with the HPLC conditions to be used. The choice of DMPK card depends on many factors such as the analyte chemical structure, extraction solvent and analysis workflow.

Specifications

Format Card
Formulation Type C
For Use With (Application) For protein-based biomolecules, on-substrate cleanup has potential to convey greater analyte stability, requires fewer experimental animals
Includes 100 cards
Key Functions Contains pure cellulose. No impregnated chemicals to interfere with analysis. Proteins are not denatured so cards may be better suited for protein based biomolecules and antibody studies.
Product Type QIAcard FTA DMPK-C