missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Carbon-Cap Disposable Activated Carbon Capsules

Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.

$345.39 - $346.26

Specifications

For Use With (Application) For water, chemical, and reagent purification
Housing Material Polypropylene
Max. Pressure 4.1 bar (60 psi)
View More Specs

Products 2
Catalog Number Mfr. No. Filtration Area Price Quantity  
Catalog Number Mfr. No. Filtration Area Price Quantity  
09-744-37
View Documents
Cytiva
67041500
82,000 m2
Each for $345.39
Only null left
 
09-744-40
View Documents
Cytiva
67047500
26000 m2
Each for $346.26
Only null left
 
Description

Description

Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.

  • Remove organic odors, color, and colloids from water and air
  • Activated carbon plus a pleated HEPA filter media retains 99.97% of all particles 0.3μm
  • Maximum housing pressure: 4.1 bar (60psi.)
  • Carbon-Cap 75 offers 26,000m2 activated carbon surface area
  • Carbon-Cap 150 offers 82,000m2 filter area equivalent

Applications:

For vacuum pump outlet removal of smoke and oil.

Specifications

Specifications

For water, chemical, and reagent purification
Polypropylene
4.1 bar (60 psi)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'