missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C

Catalog No. p-108920
Change view
Click to view available options
Diameter (Metric):
21 mm
24 mm
25 mm
32 mm
37 mm
42.5 mm
47 mm
50 mm
55 mm
70 mm
90 mm
100 mm
110 mm
125 mm
150 mm
Packaging:
Bulk
Individually Bagged
Quantity:
100 Pieces
400 Pieces
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Packaging Quantity
0987430 21 mm Bulk 100 Pieces
0987432 24 mm Bulk 100 Pieces
0987432A 25 mm Bulk 100 Pieces
09927923 25 mm Bulk 400 Pieces
09806105 32 mm Bulk 100 Pieces
0987432B 37 mm Bulk 100 Pieces
0987434 42.5 mm Bulk 100 Pieces
0987435 47 mm Bulk 100 Pieces
09927423 50 mm Bulk 100 Pieces
0987436 55 mm Bulk 100 Pieces
0987438 70 mm Bulk 100 Pieces
0987439 90 mm Bulk 100 Pieces
09927362 100 mm Individually Bagged 100 Pieces
0980523A 100 mm Bulk 100 Pieces
0987436A 110 mm Bulk 100 Pieces
0987437 125 mm Bulk 100 Pieces
0987437A 150 mm Bulk 100 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 0987430 Supplier Cytiva Supplier No. 1822021
Only null left

Combines fine particle retention with good flow rate. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C are ideal for the collection of suspended solids in potable water, natural and industrial wastes.

Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.

Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.

  • Medium to fast, high loading
  • Airflow: 6.7s/100mL
  • Air Monitoring
  • Clarification of aqueous liquids containing low to medium levels of fine particulates
  • Cell harvesting
  • Liquid scintillation counting
  • Marine chemistry and biochemistry
  • Radioimmunoassay procedures

Specifications

Basis Weight 53 g/m2
Description GE’s Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis.
Format Disc
Material Borosilicate Glass
Particle Retention 1.2 μm
Flow Rate 10.5 sec./100 mL
For Use With (Application) Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is
Grade Grade GF/C
Packaging Bulk
Quantity 100 Pieces
Shape Circle
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.01 in.
Thickness (Metric) 260 μm
Type Filter
Waterflow Rate 105 mL/min
Diameter (English) 0.82 in.
Diameter (Metric) 21 mm
Show More Show Less