Learn More
Cytiva Whatman™ Antibiotic Assay Discs
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
$463.01 - $782.44
Specifications
Product Type Antibiotic Assay Discs
Test Type Infectious Disease, Antibiotic Susceptibility Sensitivity
Packaging Type 1000 Discs/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
05712
View Documents Promotion Details
Cytiva
2017013
13 mm
Pack of 1000 Each for $782.44
Only null left
05711
View Documents Promotion Details
Cytiva
2017006
6 mm
Pack of 1000 Each for $463.01
Only null left
Description
Description
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
  • For use with Kirby-Bauer method of antibiotic testing
  • Allows rational and selective chemotherapy
Specifications
Specifications
Antibiotic Assay Discs
Infectious Disease, Antibiotic Susceptibility Sensitivity
1000 Discs/Pk.