Learn More
Cytiva Whatman™ Antibiotic Assay Discs
Description
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
- For use with Kirby-Bauer method of antibiotic testing
- Allows rational and selective chemotherapy
Specifications
Specifications
| Product Type | Antibiotic Assay Discs |
| Test Type | Infectious Disease, Antibiotic Susceptibility Sensitivity |
| Packaging Type | 1000 Discs/Pk. |
| Quantity | 1000 Discs/Pk. |
| Diameter (Metric) | 6 mm |