missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Anodisc™ Filter Membranes

Catalog No. 0992629
Change view
Click to view available options
Pore Size:
0.02 μm
0.1 μm
0.2 μm
Diameter (Metric):
13 mm
25 mm
47 mm
Includes:
No Support Ring
Support Ring
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric) Includes
0992629 0.2 μm 13 mm No Support Ring
0992630 0.1 μm 13 mm No Support Ring
0992631 0.02 μm 13 mm No Support Ring
0992633 0.1 μm 25 mm Support Ring
0992634 0.02 μm 25 mm Support Ring
0992632 0.2 μm 25 mm Support Ring
0992635 0.2 μm 47 mm Support Ring
0992636 0.1 μm 47 mm Support Ring
0992637 0.02 μm 47 mm Support Ring
09926300 0.2 μm 47 mm No Support Ring
09924567 0.02 μm 47 mm No Support Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 0992629 Supplier Cytiva Supplier No. 68097023
Only null left

Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.

Anopore inorganic membrane circles available with or without support ring contain no lateral crossovers between individual pores.
  • Wide solvent compatibility reduces the need to stock a variety of membranes in the laboratory.
  • No additives used in the manufacturing process ensures minimal extractables and no sample contamination.
  • Extremely low protein binding minimizes sample loss.
  • Virtually transparent when wet making it highly suitable for microscopy studies.

Specifications

Autoclavable Autoclavable
Material Aluminum Oxide
Pore Size 0.2 μm
Hydrophobicity Hydrophilic
Includes No Support Ring
Membrane Type Anodisc 13
Quantity 100 Pack
Diameter (Metric) 13 mm