Learn More
Cytiva Whatman™ Acid/Alkali Test Papers
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
$35.98 - $177.19
Specifications
pH Range 5 to 8
Type pH Test Paper
View More Specs
Products 3
Catalog Number Mfr. No. Color Indicator Format Price Quantity  
Catalog Number Mfr. No. Color Indicator Format Price Quantity  
09-876-6
View Documents
Cytiva
2600601
Litmus Blue Strip
Pack of 20 Each for $177.19
Only null left
09927821
View Documents
Cytiva
2600202A
Litmus Red Reel
Each for $35.98
Only null left
09-876-7
View Documents
Cytiva
2600602
Litmus Red Strip
Pack of 20 Each for $177.19
Only null left
Description
Description
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
  • Litmus Blue turns red in acid
  • Litmus Red turns blue in base, at pH 5 to 8
  • Congo Red ph range: 6.8 to 8.3
Order Info
20 leaves per book. 10 x 10 books per pack.
Specifications
Specifications
5 to 8
pH Test Paper
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'