missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Acid/Alkali Test Papers

Catalog No. 098766
Change view
Click to view available options
Color Indicator:
Litmus Blue
Litmus Red
Format:
Reel
Strip
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Indicator Format
098766 Litmus Blue Strip
09927821 Litmus Red Reel
098767 Litmus Red Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 098766 Supplier Cytiva Supplier No. 2600601
Only null left

Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.

  • Litmus Blue turns red in acid
  • Litmus Red turns blue in base, at pH 5 to 8
  • Congo Red ph range: 6.8 to 8.3

Order Info

20 leaves per book. 10 x 10 books per pack.

Specifications

Color Indicator Litmus Blue
pH Range 5 to 8
Format Strip
Quantity 10 x 10 Books of 20 Strips
Type pH Test Paper
Length (Metric) 60 mm
Width (Metric) 10 mm