missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 3 μm Pore Size (ME 29) Mixed Cellulose Ester Membranes, Plain
Whatman™ (ME 29) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.
Supplier: Cytiva 10400712
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 29
- Filter Type: Plain
- Pore Size: 3 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Bubble Point: 1.3 bar
- Flow Rate: 1.2 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 6.7 sec./100 mL (at Δp = 3 mbar)
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
| Autoclavable | |
| ME 29 | |
| Non-sterile | |
| Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
| Plain | |
| 47 mm |
| MCE | |
| 3 μm | |
| 1.2 sec./100 mL/12.5 cm² (at Δp = 0.9 bar) | |
| 6.7 sec./100 mL (at Δp = 3 mbar) | |
| 1.85 in. |