Learn More
Cytiva Whatman™ 1PS Disposable Phase Separating Paper
Creates a hydrophobic filter that can replace separatory funnel in some applications. Whatman™ 1PS Disposable Phase Separating Paper is impregnated with stable silicone to allow solvent phases to pass through while retaining aqueous phases.
Supplier:  Cytiva 2200070
 View more versions of this product
Catalog No. 09800975
Only null left
Description
Description
Creates a hydrophobic filter that can replace separatory funnel in some applications. Whatman™ 1PS Disposable Phase Separating Paper is impregnated with stable silicone to allow solvent phases to pass through while retaining aqueous phases.
Passes organic solvent phases lighter or heavier than water but retains water phase.
  • Hydrophobic
  • Use folded in conical funnel
  • Disposable
  • Eliminates need for washing and drying separatory funnels, cross-contamination
Order Info
In 7 to 24cm diameter circles.
Specifications
Chemical Identifiers
Fiberglass/Silicone
Liquid phase separation
Separator Paper
Specifications
Fiberglass/Silicone
100Pack
2.75 in.
Liquid phase separation
Separator Paper
70 mm
Product Suggestions
Product Suggestions