missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Latex Palm

Fit and comfortable protection

$118.04 - $118.04

Specifications

Color Gray
Cut Resistance Rating ANSI A3
Material Latex, Natural Rubber, Foam
Product Type Cut-resistant Gloves
For Use With (Application) Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
View More Specs

Alert:

This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 5
Catalog Number Mfr. No. Glove Cuff Color Size For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Glove Cuff Color Size For Use With (Application) Price Quantity  
19148741
View Documents
Wells Lamont™
WLCY9282S
Red Small Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Dozen of 12 for $118.04
Case of 12 Dozen for $1,285.80
Only null left
 
19148742
View Documents
Wells Lamont™
WLCY9282M
Green Medium Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Dozen of 12 for $118.04
Case of 12 Dozen for $1,285.80
Only null left
 
19148733
View Documents
Wells Lamont™
WLCY9282L
Brown Large Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Dozen of 12 for $118.04
Case of 12 Dozen for $1,285.80
Only null left
 
19148734
View Documents
Wells Lamont™
WLCY9282XL
Blue X-Large Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Dozen of 12 for $118.04
Case of 12 Dozen for $1,285.80
Only null left
 
19148762
View Documents
Wells Lamont™
Y9282XXL
Black 2X-Large Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Dozen of 12 for $118.04
Case of 12 Dozen for $1,211.28
Only null left
 
Description

Description

  • Features highly cut-resistant fiber for protection against lacerations
  • Foam-latex palm coating improves grip
  • Knit wrist construction prevents dirt and debris from entering the glove
  • Color: Gray

Specifications

Specifications

Gray
Latex, Natural Rubber, Foam
Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
ANSI A3
Cut-resistant Gloves
Knit
Videos
SDS
Documents

Documents