Learn More
Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Latex Palm
Fit and comfortable protection
Supplier: Wells Lamont™ Y9282XXL
Alert:
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
- Features highly cut-resistant fiber for protection against lacerations
- Foam-latex palm coating improves grip
- Knit wrist construction prevents dirt and debris from entering the glove
- Color: Gray
Specifications
| Gray | |
| Latex, Natural Rubber, Foam | |
| 2X-Large | |
| Black |
| ANSI A3 | |
| Cut-resistant Gloves | |
| Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove | |
| Knit |