Learn More
Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Latex Palm
Fit and comfortable protection
Supplier:  Wells Lamont™ WLCY9282L
 View more versions of this product
Catalog No. 19148733
Only null left
Explore available promotions
Alert:
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
  • Features highly cut-resistant fiber for protection against lacerations
  • Foam-latex palm coating improves grip
  • Knit wrist construction prevents dirt and debris from entering the glove
  • Color: Gray
Specifications
Specifications
Gray
Latex, Natural Rubber, Foam
Large
Brown
ANSI A3
Cut-resistant Gloves
Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Knit
Product Suggestions
Product Suggestions