missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Grain Cowhide Leather Drivers Gloves with Keystone Thumb

Grain Cowhide Leather Drivers Gloves are constructed from high quality materials to provide comfort and protection while on the job. A shirred elastic wrist prevents particles from entering the glove, while a keystone thumb adds movement and comfort.

$63.70 - $162.95

Specifications

Back Material Shirred Elastic
Color Natural
Grade Economy
Hand Specific
Material Grain Cowhide Leather
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19823914
View Documents
Wells Lamont™
Y0131S
Small
Dozen of 12 for $63.70
Case of 12 Dozen for $655.20
Only null left
 
19823915
View Documents
Wells Lamont™
Y0131M
Medium
Dozen of 12 for $63.70
Case of 12 Dozen for $655.20
Only null left
 
19823916
View Documents
Wells Lamont™
Y0131L
Large
Dozen of 12 for $137.54
Case of 12 Dozen for $1,414.68
Only null left
 
19823917
View Documents
Wells Lamont™
Y0131XL
X-Large
Dozen of 12 for $67.68
Case of 12 Dozen for $696.24
Only null left
 
19148795
View Documents
Wells Lamont™
Y0131XXL
2X-Large
Dozen of 12 Pair for $162.95
Case of 6 Dozen for $888.84
Only null left
 
Description

Description

  • Economy option
  • Elastic shirred back
  • Bound hem
  • Keystone thumb
Specifications

Specifications

Shirred Elastic
Economy
Grain Cowhide Leather
Landscaping, Farming, Warehousing, Construction, Oil and Gas Industries
Y0131
Natural
Specific
Driver Gloves
Open
Keystone
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'