Learn More
Wells Lamont™ Grain Cowhide Leather Drivers Gloves with Keystone Thumb
Grain Cowhide Leather Drivers Gloves are constructed from high quality materials to provide comfort and protection while on the job. A shirred elastic wrist prevents particles from entering the glove, while a keystone thumb adds movement and comfort.
Supplier: Wells Lamont™ Y0131XXL
Description
- Economy option
- Elastic shirred back
- Bound hem
- Keystone thumb
Specifications
| Shirred Elastic | |
| Economy | |
| Grain Cowhide Leather | |
| 2X-Large | |
| Open | |
| Y0131 |
| Natural | |
| Specific | |
| Driver Gloves | |
| Landscaping, Farming, Warehousing, Construction, Oil and Gas Industries | |
| Black | |
| Keystone |