missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Grain Cowhide Leather Drivers Gloves with Keystone Thumb

Catalog No. 19823917
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19823917 X-Large
19823914 Small
19823915 Medium
19823916 Large
19148795 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19823917 Supplier Wells Lamont™ Supplier No. Y0131XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Grain Cowhide Leather Drivers Gloves are constructed from high quality materials to provide comfort and protection while on the job. A shirred elastic wrist prevents particles from entering the glove, while a keystone thumb adds movement and comfort.

  • Economy option
  • Elastic shirred back
  • Bound hem
  • Keystone thumb

Specifications

Back Material Shirred Elastic
Color Natural
Grade Economy
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size X-Large
For Use With (Application) Landscaping, Farming, Warehousing, Construction, Oil and Gas Industries
Gender Unisex
Glove Cuff Color Blue
Glove Cuff Style Open
Hem Color Blue
Series Y0131
Thumb Style Keystone
Show More Show Less