missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WAY 316606 hydrochloride

Catalog No. 476710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
476710 10 mg
476750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 476710 Supplier Tocris Bioscience™ Supplier No. 4767/10
Only null left

Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling

WAY 316606 hydrochloride is a secreted Frizzled-Related Protein-1 (sFRP-1) inhibitor (IC50 = 0.65 μM); prevents sFRP-1 from interacting with Wnt and thus increases Wnt signaling. Increases total bone area in a murine calvarial organ culture assay.

We are aware of a study in PLOS Biology where WAY-316606 was found to stimulate hair follicle growth. Please note that WAY-316606 from Tocris is for laboratory research use only, and that we will only sell to established scientific businesses and institutes.

Specifications

CAS 1781835-02-6
Quantity 10 mg
Product Type WAY 316606 hydrochloride
Purity >99%
Molecular Weight (g/mol) 484.94
Inhibitors WAY 316606 hydrochloride

For Research Use Only