missing translation for 'onlineSavingsMsg'
Learn More

Water, HPLC Grade, Spectrum™ Chemical

$376.50 - $376.50

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614669
View Documents
Spectrum Chemical Mfg Cor
HP9024LT
4 L Amber Glass Bottle
Each for $376.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

7732-18-5
High Purity Solvent
1 ppm
1
Amber Glass Bottle
4 L
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.