missing translation for 'onlineSavingsMsg'
Learn More

Water, HPLC Grade, Spectrum™ Chemical

Catalog No. 18614669
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614669 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614669 Supplier Spectrum Chemical Mfg Cor Supplier No. HP9024LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 18.02

Specifications

CAS Max % 1
Grade HPLC
Packaging Amber Glass Bottle
Residue after Evaporation 1 ppm
Sterility Non-sterile
Quantity 4 L
Chemical Name or Material Water

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.