Learn More
Water for HPCE, For Luminescence and UV-Spectroscopy, MilliporeSigma™ Supelco™
Water is widely used for HPCE (High-Performance Capillary Electrophoresis), luminescence and UV-spectroscopic studies.
$64.22 - $216.73
Chemical Identifiers
MDL Number MFCD00011332
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015494
View Documents
Merck Emd Millipore
95283100ML
100 mL
Each for $64.22
Only null left
111015498
View Documents
Merck Emd Millipore
952831L
1 L
Each for $216.73
Only null left
Description
Description
Specifications
Chemical Identifiers
MFCD00011332
Specifications
7732-18-5
100°C
MFCD00011332
18.02
1.000 g/mL (at 3.98°C)
H2O
100 mL
For HPCE
Videos
Safety and Handling
Safety and Handling
RTECSNumber : ZC0110000
SDS
missing translation for 'documents'
missing translation for 'documents'