missing translation for 'onlineSavingsMsg'
Learn More

Water for HPCE, For Luminescence and UV-Spectroscopy, MilliporeSigma™ Supelco™

Catalog No. 111015494
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015494 100 mL
111015498 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015494 Supplier Merck Emd Millipore Supplier No. 95283100ML

Water is widely used for HPCE (High-Performance Capillary Electrophoresis), luminescence and UV-spectroscopic studies.

Chemical Identifiers

MDL Number MFCD00011332

Specifications

Density 1.000 g/mL (at 3.98°C)
CAS 7732-18-5
Boiling Point 100°C
Linear Formula H2O
Quantity 100 mL
Formula Weight 18.02
Grade For HPCE
RTECSNumber ZC0110000