missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Hardness Buffer, Odorless, Ricca Chemical
$128.27 - $576.23
Specifications
| Chemical Name or Material | Water Hardness Buffer |
|---|---|
| Name Note | Odorless |
| CAS | 7732-18-5,141-43-5,7647-01-0,6381-92-6,7791-18-6 |
| CAS Min % | 71.0,21.62,4.62,0.34,0.18 |
| CAS Max % | 74.0,22.5,4.81,0.35,0.19 |
Specifications
| Water Hardness Buffer | |
| 7732-18-5,141-43-5,7647-01-0,6381-92-6,7791-18-6 | |
| 74.0,22.5,4.81,0.35,0.19 | |
| 1X | |
| Liquid | |
| Natural Poly Bottle |
| Odorless | |
| 71.0,21.62,4.62,0.34,0.18 | |
| Colorless to Yellow | |
| Water Analysis | |
| Buffer | |
| Laboratory |