missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Hardness Buffer, Odorless, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Water Hardness Buffer |
| Name Note | Odorless |
| CAS | 7732-18-5, 141-43-5, 7647-01-0, 6381-92-6, 7791-18-6 |
| CAS Min % | 71.0, 21.62, 4.62, 0.34, 0.18 |
| Color | Colorless to Yellow |
| CAS Max % | 74.0, 22.5, 4.81, 0.35, 0.19 |
| Concentration | 1X |
| For Use With (Application) | Water Analysis |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Show More |