missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, For ion chromatography, MilliporeSigma™ Supelco™
Water is of high purity and is used especially for the needs of ion chromatography (IC). It is suitable for trace analysis of anions, cations and some organics, which are usually analyzed by ion chromatography (IC).
$98.58 - $163.57
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| MDL Number | MFCD00011332 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| IUPAC Name | water |
| SMILES | O |