missing translation for 'onlineSavingsMsg'
Learn More

Water, For ion chromatography, MilliporeSigma™ Supelco™

Catalog No. 111015499
Change view
Click to view available options
Quantity:
2.5 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015499 2.5 L
111015500 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015499 Supplier Merck Emd Millipore Supplier No. 006122.5L

Water is of high purity and is used especially for the needs of ion chromatography (IC). It is suitable for trace analysis of anions, cations and some organics, which are usually analyzed by ion chromatography (IC).

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Linear Formula H2O
Formula Weight 18.02
Grade For ion chromatography
Density 1.000 g/mL (at 3.98°C)
Quantity 2.5 L
Refractive Index n20/D 1.34
RTECSNumber ZC0110000